Haloacetic Acid Mix (9 Components), Restek
Supplier: Restek
31896
75834-868EA 114.74 USD
75834-868
Haloacetic Acid Mix (9 Components), Restek
Standards Organic Standards
This consists of bromochloroacetic acid, bromodichloroacetic acid, chlorodibromoacetic acid, dibromoacetic acid, dichloroacetic acid, monobromoacetic acid, monochloroacetic acid, tribromoacetic acid and trichloroacetic acid.