Phenylurea Pesticide Mixture (8 Components), Restek
Supplier: Restek
32434
75834-992EA 69.39 USD
75834-992
Phenylurea Pesticide Mixture (8 Components), Restek
Standards Organic Standards
Contains eight components, those are diflubenzuron, diuron, fluometuron, linuron, propanil, siduron, tebuthiuron and thidiazuron.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.