Ketones Mix, 524.2 Rev. 4.1 (5 Components), Restek
Supplier: Restek
30602
75835-588EA 63.3 USD
75835-588
Ketones Mix, 524.2 Rev. 4.1 (5 Components), Restek
Standards Organic Standards
The ketones mix consists of five components they are acetone, 2-butanone (MEK), 1,1-dichloro-2-propanone, 2-hexanone and 4-methyl-2-pentanone (MIBK).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.