508.1 Calibration Mix #2 (11 Components), Restek
Supplier: Restek
32095
75834-224EA 80.26 USD
75834-224
508.1 Calibration Mix #2 (11 Components), Restek
Standards Organic Standards
Consists of 11 components, those are chlorobenzilate, cis-chlordane, trans-chlordane, chloroneb, dcpa methyl ester , etridiazole, hexachlorobenzene, cis-permethrin*, trans-permethrin*, propachlor and trifluralin.
11 components, Contains 500 µg/mL total permethrin.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Ordering information: Exact content of each isomer is listed on certificate of analysis.