D-(+)-Biotin

Synonyms: Vitamin HVitamin B85-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acidD-BiotinVitamin B7

9587-10G 9587-5G
75837-468EA 341.61 USD
75837-468 75837-466
D-(+)-Biotin
D(+)-Biotin

D-(+)-Biotin is a coenzyme and a B vitamin. May be used to elute proteins from avidin/streptavidin resins.

Formula: C₁₀H₁₆N₂O₃S
MW: 244.31 g/mol
MDL Number: MFCD00005541
CAS Number: 58-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR