Azathioprine

Synonyms: 6-(1-Methyl-4-nitroimidazol-5-yl)thiopurine

9536-100 9536-500
75837-244EA 64.21 USD
75837-244 75837-246
Azathioprine
Azathioprine

Azathioprine is a purine analog that displays immunosuppressive and anti-inflammatory properties. It serves as a prodrug for 6-mercaptopurine, which alters purine processing and DNA synthesis, interfering with the proliferation of lymphocytes and other myeloid cells. 

Formula: C₉H₇N₇O₂S
MW: 277.26 g/mol
MDL Number: MFCD00069203
CAS Number: 446-86-6
UN: 3316
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR