Glycerine ≥99.5%

Supplier: AQUA SOLUTIONS MS

Synonyms: GlysaninGlycerinGlyceritolPropane-1,2,3-triolOsmoglynPropanetriolGlycerolGlyrolGlycyl alcohol1,2,3-PropanetriolTrihydroxypropane

G7502-4L
101224-392EA 253.19 USD
101224-392
Glycerine ≥99.5%
Glycerine
Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 182 °C
Melting Pt: 20 °C
Density: 1.26 g/cm³(20 °C)
Flash Pt: 160 °C
MDL Number: MFCD00004722
CAS Number: 56-81-5
https://us.vwr.com/stibo/web/std.lang.all/47/74/734774.jpg

Order Now

Specification Test Results

Solubility in water Fully miscible

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR