Retinoic acid, Powder USP

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: all-trans-Retinoic acidVitamin A acidATRATretinoin

R1022-100GMB R1022-1KGBL R1022-5GM R1022-25GM R1022-1GM R1022-1GM-CS12
700009-308EA 4208.32 USD
700009-308 75812-112 700009-310 700009-312 700009-314 700009-316
Retinoic acid, Powder USP
Retinoic acid

Tretinoin, Powder, USP is used to treat acne. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: C₂₀H₂₈O₂
Melting Pt: 180…182 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001551
CAS Number: 302-79-4

Order Now

Specification Test Results

Assay (Dried Basis) 97.0 - 103.0%
Loss on Drying 0.50%
Residue on Ignition 0.10%
Isotretinoin 5.00%
Heavy Metals 0.002%
Identification Passes Test
Residual Solvents Passes Test
Partition Coefficient (n-Octanol/Water) 6.7
Solubility Insoluble in Water and Soluble in Diethyl Ether

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR