Would you like to log in again?
Ward's® Aquatic D Frame Net
Supplier: VWR
100625 P24343
Effectively Samples Shallow Bottom Sediments
  • Easy to use
  • Sturdy reinforced build
The unique steel design of this 12" frame makes it easier to sweep over aquatic vegetation, shallow pond bottoms, or near the shore. The handle has a reinforced frame, and the bag is constructed of 1200 µm mesh.
Log in to see your contract pricing and availability.
  Remember me on this device
Expand All / Collapse All
Description Depth Handle Length Width Supplier No. VWR Catalog Number Unit Price Quantity
Ward's® Aquatic Net 18" 48" 12"   470220-356 Kit of 1 Retrieving
Replacement Parts
Description Depth Width Supplier No. VWR Catalog Number Unit Price Quantity
Replacement Net 18" 12" P24343 470018-508 Each Retrieving
Replacement Handle - - 100625 470018-514 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty