3-Fluoro-N-methylbenzylamine ≥97%

Danger

Synonyms: 1-(3-Fluorophenyl)-N-methylmethanamine(3-Fluorobenzyl)methylamine

H56177-14 H56177-06 H56177-06 H56177-14
AAH56177-06EA 137.32 USD
CAAAH56177-14 CAAAH56177-06 AAH56177-06 AAH56177-14
3-Fluoro-N-methylbenzylamine ≥97%
3-Fluoro-N-methylbenzylamine
Formula: C₈H₁₀FN
MW: 139.17 g/mol
Boiling Pt: 183…184 °C
Density: 1,015
Flash Pt: 66 °C (151 °F)
Storage Temperature: Ambient
MDL Number: MFCD04623547
CAS Number: 90389-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR