Apa I (Blue/White Cloning Qualified)
Supplier: Promega
R6361
PAR6361EA 131.38 USD
PAR6361
Apa I (Blue/White Cloning Qualified)
Nucleic Acid Reagents Restriction Enzymes
G GGCC/C C\CCGG G
  • Active and capable of digestion directly in GoTaq(R) Green Master Mix
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications