Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ToxOut™ Endotoxin Free PBS, BioVision

Supplier: BioVision
7943-50
50 ml of PBS recommended to use for samples with special requirement of endotoxin level.

  • Recommended to use for samples with special requirement of endotoxin level
  • Isotonic and non-toxic to cells, Maintains osmolarity
  • Endotoxin Free PBS

Endotoxin is the lipopolysaccharide (LPS) complex located in the outer membrane of gram-negative bacteria. During experimental procedures, large amount of endotoxins are shed and can easily contaminate labware, buffers and downstream products. Presence of endotoxin in any of these has a detrimental effect on culturing of cells and can lead to generation of irreproducible or negative experimental data. Therefore, it is critical to keep endotoxin away from samples and products. ToxOut™ Endotoxin-free PBS is recommended to use for samples with special requirement of endotoxin level (< 0.05 EU/ml). PBS is one of the most commonly used buffers, which is isotonic and non-toxic to cells and has the ability to maintain their osmolarity, thereby making the buffer suitable for washing procedures in cell cultures and for immunoassays.




 

ORDER

Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
ToxOut™ Endotoxin-Free PBS 7943-50 75838-190 Each (50ml) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty