EZAct™ Procathepsin Activation Buffer, BioVision
Supplier: BioVision
1030-1
75838-172EA 134.26 USD
75838-172
EZAct™ Procathepsin Activation Buffer, BioVision
Buffers Biological Buffers
A ready-to-use buffer for generation of active cysteine/aspartyl Cathepsins by auto-activation of the Procathepsins.
  • To generate active Cathepsins from inactive Procathepsins.
  • To test the activity of Cathepsins.
  • To screen for Cathepsin Inhibitor candidates.
Cathepsins (CTS) are proteases that degrade proteins. They are present in all animals and other organisms. These proteases are of three types: cysteine proteases, aspartyl protease and serine proteases. Cathepsins are produced as inactive proenzyme precursors that are subsequently activated in lysosomes. Biovision’s EZAct™ Procathepsin Activation Buffer is a ready-to-use buffer for generation of active cysteine/aspartyl Cathepsins by auto-activation of the Procathepsins.