SP Wilmad-LabGlass Reaction Kettle Clamps, SP Industries
Supplier: SP Industries
LG-7317-102 LG-7317-100
14200-842EA 286.99 USD
14200-842 14201-230
SP Wilmad-LabGlass Reaction Kettle Clamps, SP Industries
Clamps General Purpose Clamps
These reaction vessel clamps for O-Rings and flat flanges are a cast bronze ring with a black oxidized finish
Three insulated, spring activated clamping arms fit beneath the flange of the flask body. Simple pressure on the heat-resistant fiber knobs, and a twist, enable attachment or removal.