Would you like to log in again?
Double Vacuum Manifold, Wilmad-LabGlass
Supplier: WILMAD/LABGLASS
IG-357-100
14207-576EA 2221.4 USD InStock
14207-576
Double Vacuum Manifold, Wilmad-LabGlass
Manifolds
This borosilicate glass air-tite vacuum gas manifold (schlenk line) is designed with double oblique pressure/vacuum stopcocks with 4 mm bores and a 16.2/56 plug size
The stopcocks simplify evacuation or pressurization with an inert atmosphere. Manifold is 1" O.D.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty