SP Wilmad-LabGlass Universal Inlet Adapters with Screw Cap, SP Industries
Supplier: SP Industries
LG-1451-404 LG-1451-304 LG-1451-400 LG-1451-200 LG-1451-102 LG-1451-090 LG-1451-308 LG-1451-406 LG-1451-306 LG-1451-100 LG-1451-108 LG-1451-300 LG-1451-104 LG-1451-204
82002-994EA 48.76 USD
82002-994 14211-346 14211-246 14211-420 14204-408 14201-560 14209-206 14204-402 14202-038 14204-404 14210-496 14202-034 14212-786 14202-494
SP Wilmad-LabGlass Universal Inlet Adapters with Screw Cap, SP Industries
Adapters Adapters, Conical Ground Joints
Borosilicate glass universal inlet adapters are capable of sealing thermometers, syringes, pipets, and more.
  • Top screw cap
  • Bottom [ST] joint
  • Screw cap and o-ring included
Insert probes can be positioned within the apparatus by loosening the screw cap on top of adapter and tightening to create a seal. The bottom of the adapter features a Standard Taper joint. Supplied as a complete assembly with a screw cap and Viton® O-ring.