SP Wilmad-LabGlass Solubility Tubes, SP Industries
Supplier: SP Industries
LG-10700-102
14007-632EA 51.11 USD
14007-632
SP Wilmad-LabGlass Solubility Tubes, SP Industries
Tubes Test Tubes
These borosilicate glass solubility tubes are available with or without constriction.
  • Strong
  • Durable
They are designed to find the solubility index, and can be used in a variety of laboratory testing analysis.