Trypticase™ Soy Agar with Penicillinase, BD Diagnostics
Supplier: BD Diagnostics
221839
90006-494PK 129.44 USD
90006-494
Trypticase™ Soy Agar with Penicillinase, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Trypticase™ Soy Agar with Penicillinase is recommended for the detection and enumeration of microorganisms present on surfaces of sanitary importance.
  • Penicillinase inactivates antibiotics such as penicillins and cephalosporins
  • Double-wrapped
  • Sterilized via gamma radiation to allow removal without contamination
  • Trustworthy microbial growth due to post-packaging sterilization
Since the agar medium has been sterilized after packaging, the presence of microbial growth after sampling and incubation can be relied upon to represent true recovery and not pre-existing medium contaminants.