BBL™ Modified mTEC Agar, BD
Supplier: BD Life Sciences
215044 215044
90006-314PK 482.67 USD
CA90006-314 90006-314
BBL™ Modified mTEC Agar, BD
Media for Microbiology Prepared Media for Microbiology
Modified mTEC Agar is a selective culture medium used for the chromogenic detection and enumeration of thermotolerant Escherichia coli in water by the membrane filtration technique.
  • Peptone is a source of nitrogen, carbon, and amino acids
  • Yeast extract provides trace elements, vitamins, and amino acids
  • Lactose is a fermentable carbohydrate and carbon source
  • Sodium chloride maintains osmotic equilibrium
  • Agar is the solidifying agent
Red or magenta colonies may be presumptively identified as E. coli.