BBL™ Mueller Hinton II Agar, BD
Supplier: BD Life Sciences
221177 221177 221275 221275
90006-210PK 175.48 USD
90006-210 CA90006-180 CA-90006-210 90006-180
BBL™ Mueller Hinton II Agar, BD
Media for Microbiology Prepared Media for Microbiology
Mueller Hinton II Agar is used in the standardized disc diffusion procedure for determining the susceptibility of rapidly growing aerobic organisms to antimicrobial agents.
  • Digest of casein and beef extract supply essential nutrients
  • Starch acts as a protective colloid against toxic substances
  • Produced dextrose is a source of energy
  • Agar is the solidifying agent
Mueller Hinton II Agar complies with requirements of the World Health Organization and is specified in the FDA Bacteriological Analytical Manual for food testing.