Would you like to log in again?
UCH-L1 Deubiquitinase Inhibitor Screening Kit, BioVision Inc.
Supplier: BioVision
K484-100
Validated using UCH-L1 inhibitor.
  • Rapid, simple & reliable test for screening potential inhibitors of UCH-L1
  • Includes UCH-L1 Inhibitor [LDN-57444]
Ubiquitin C-terminal Hydrolase 1 (UCH-L1) is a deubiquitinating enzyme (DUB) that is expressed predominantly in neuronal tissue, comprising up to 1-2% of total protein in some brain tissues. UCH-L1 is a cysteine protease and capable of cleaving the isopeptide bond between the carboxyl end of an ubiquitin molecule and a lysine residue on the ubiquitin-modified protein. As ubiquitination is utilized for both localization and degradation of these modified proteins, proper function of deubiquinating enzymes is essential for cell viability and integrity. Dysregulation of UCH-L1 has been implicated in the pathophysiology of neurological disorders like Parkinson’s and Alzheimer’s diseases, as well as in cancer invasion. A further understanding of these processes can be facilitated by identification of inhibitors with high selectivity. With over 400 DUB enzymes known, this target offers far more selectivity than the proteasome system. BioVision’s UCH-L1 Inhibitor Screening Kit utilizes the ability of active UCH-L1 deubiquitinase to cleave a synthetic protein substrate to release the free fluorophore, which can be easily quantified (Ex/Em = 350/440 nm) using a fluorescence microplate reader. Small molecule inhibitors can either reduce or abolish this activity. This inhibitor screening kit thus allows rapid and reliable determination of the inhibitory effects of various compounds on UCH-L1 deubiquitinase and should be used to screen for novel inhibitors.
ORDER
Expand All / Collapse All
Description Size Ex/Em Supplier No. VWR Catalog Number Unit Price Quantity
UCH-L1 Inhibitor Screening Kit (Fluorometric) 100 Assays 350/440 nm K484-100 75836-740 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Detection Method Fluorescence
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty