Would you like to log in again?
Angiotensin I Converting Enzyme Activity (ACE1) Assay Kit (Fluorometric), BioVision Inc.
Supplier: BioVision
K227-100
Industry's first kit to detect ACE1 activity in biological samples.
  • Simple one-step reaction
  • Takes only one to two hours
  • Non-radiometric fluorescent detection
  • HTP adaptable
Angiotensin I converting enzyme (ACE1, EC: 3.4.15.1), a dipeptidyl carboxypeptidase, is part of the renin-angiotensin system (RAS) that controls regulation of blood pressure by cleaving the C-terminal dipeptides of angiotensin I and bradykinin. It is found on the luminal surface of vascular endothelial cells, especially in pulmonary tissues. Elevated levels of ACE1 are found in patients suffering from sarcoidosis, leprosy, hyperthyroidism, acute hepatitis, primary biliary cirrhosis, diabetes mellitus, multiple myeloma, osteoarthritis, amyloidosis, Gaucher’s disease, pneumoconiosis, histoplasmosis and miliary tuberculosis. BioVision’s ACE1 Activity Assay Kit utilizes an active ACE1 to cleave a synthetic o-aminobenzoyl peptide (Abz-based peptide) substrate to release a fluorophore. The released Abz can be easily quantified using a fluorescence microplate reader. This assay kit is simple, rapid and can detect ACE activity as low as 10 mU in biological samples.
ORDER
Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
Angiotensin I Converting Enzyme Activity (ACE) Assay Kit (Fluorometric) 100 Assays K227-100 75836-724 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty