Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ExoStd™ Lyophilized Exosome Standard, 100 µg, MM1 Cell Line, BioVision Inc

Supplier: BioVision
M1051-4 M1051-2 M1051-6
Lyophilized and purified Exosomes available from MM1 cell line (100 µg).

  • Purified and lyophilized exosomes
  • Control standards for multiple applications including FACS, WB, ELISA
  • Calibration standards for quantitation of exosome-derived markers from biological samples
  • Easy to ship and store
  • Stable for over 36 months at 4°C




 

ORDER

Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
Lyophilized Exosome Standard (100 µg, MM1 cell line) 2 vials M1051-2 75799-606 Each Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

Lyophilized Exosome Standard (100 µg, MM1 cell line) 4 vials M1051-4 75799-608 Each Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

Lyophilized Exosome Standard (100 µg, MM1 cell line) 6 vials M1051-6 75799-610 Each Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty