ExoStd™ Lyophilized Exosome Standard, 30 µg, Human Saliva, BioVision Inc
Supplier: BioVision
M1046-6
75799-516EA 1348.07 USD
75799-516
ExoStd™ Lyophilized Exosome Standard, 30 µg, Human Saliva, BioVision Inc
Lyophilized and purified Exosomes available from Human Saliva (30 µg).
  • Purified and lyophilized exosomes
  • Control standards for multiple applications including FACS, WB, ELISA
  • Calibration standards for quantitation of exosome-derived markers from biological samples
  • Easy to ship and store
  • Stable for over 36 months at 4°C