Brady BBP®37 Multicolor Sign and Label Printer and Kits, Brady®
Supplier: Brady Worldwide
BBP37
10017-938EA 5175.72 USD
10017-938
Brady BBP®37 Multicolor Sign and Label Printer and Kits, Brady®
Printers
Color your way to an improved workplace with the BBP®37 Color and Cut, Sign and Label Printer
  • Automatic label setup
  • 20-second material change out
  • 5-inch per second print speed
Waiting for signs and labels is a thing of the past. Create bright and colorful labels and signs that really make an impact in your facility. With 28 different industrial grade label and ribbon materials to choose from, you can print visuals for nearly any facility application. Not to mention all these materials simply click or set into place without calibration or threading. With the XY cutter, you can create die-cut shapes and text that give you the flexibility to put your messages where you need them most. The BBP37 Printer can connect to a PC but also operates as a stand-alone printer. In fact, the built in touchscreen interface and preset label templates makes label creation a breeze. Order the BBP37 Color and Cut, Sign and Label Printer today or request a demo to learn more!
Ordering information: BBP® 37 Printer; Cleaning Kit; Cutter Cleaning Tool; Power Cord; Quick Start Guide; USB Cable.