Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Listeria Antibody Kit, AOAC Approved, Hardy Diagnostics

Supplier: HARDY DIAGNOSTICS

Antibody kit for Listeria testing in most applications.

  • Antibody kit includes sufficient target antibody and negative control for 60 pathogen detection tests and their associated negative control
  • Additional confirmatory test (if required)
  • AOAC Approved for raw ground beef, beef trim and spinach
Antibody kit for Listeria testing in most applications. Target Ab bound to 3 micron microspheres. 2ml sealed glass vials with pull tab. Lyophilized stabilization at room temperature. When wet store at 2 to 8 ° C. Disruption in an aligned liquid crystal matrix within the BioCassette caused by aggregation of target pathogens binding to their associated antibodies. To be used with CDx BioCassette.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Description Size No. of Preparations Packaging Supplier No. VWR Catalog Number Unit Price Quantity
Listeria Antibody Kit 1 ml per vial
5 tests/vial
60 3 ml glass vial 112520 75838-648 Pack of 60 Retrieving
Listeria Antibody Kit 1 ml per vial
5 tests/vial
120 3 ml glass vial 124520 75838-642 Pack of 120 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antibody Type Primary
Antigen Symbol IgG
Clonality Polyclonal
Conjugation Microspheres, 2.8 μm
Host Goat
Reactivity Listeria
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty