Letheen Broth, Hardy Diagnostics
Supplier: Hardy Diagnostics
U39
89413-494PK 114.71 USD
89413-494
Letheen Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Letheen Broth to determine bactericidal activity of quaternary ammonium compounds.
For use to determine bactericidal activity of quaternary ammonium compounds.