SnapShot™ ATP Bioluminescence, Hardy Diagnostics
Supplier: Hardy Diagnostics
SPXL1333
75836-682PK 441.66 USD
75836-682
SnapShot™ ATP Bioluminescence, Hardy Diagnostics
SnapShot is designed to help companies conducting ATP monitoring with a 3M™ or Bio-Trace™ luminometer reduce costs and achieve more accurate results.
  • Patented device design enables superior sample recovery
  • Sensitive to low levels of contamination
  • More tolerant to temperature abuse and sanitizers
  • SnapShot uses less materials and plastic than other
  • 100% recyclable
SnapShot is designed to help companies conducting ATP monitoring with a 3M™ or Bio-Trace™ luminometer reduce costs and achieve more accurate results. SnapShot works exactly the same as the test device it is replacing so there is no need to adjust Pass/Fail setting in the luminometer.