Privo™ Protective Eyewear, 3M™
Supplier: 3M
7000127538
89402-960CS 224 USD
89402-960
Privo™ Protective Eyewear, 3M™
Safety Glasses
Privo™ unisex protective eyewear looks as smooth as it functions.
  • Glasses provide safety on the job, with a sleek appearance for use away from the jobsite
  • Slight wrap of the lenses looks stylish and offers extra eye protection
  • Soft nose bridge and temple tips cushion high contact points for comfortable, all-day wear
  • Impact-resistant polycarbonate lens absorbs 99.9% UV rays
3M™ Privo™ Protective Eyewear combines a sophisticated European style with the practicality of safety glasses.
Eye protection is important to safeguard the user's eyes and maintain healthy vision. According to the Bureau of Labor Statistics, nearly three out of every five workers who suffered eye injuries were not wearing eye protection at the time of the accident. Wearing the correct safety eyewear for the task and for the unique needs of the worker is important. All 3M™ protective eyewear combines safety features with comfort enhancements.
These safety glasses are recommended for use in assembly and mechanical, chipping, chiseling, drilling, grinding, machining, masonry, metal repair, painting, pouring/casting, power fastening, riveting, sanding, and sawing applications. Industries in which these safety glasses are commonly used include aviation, construction, food and beverage, general manufacturing, mining, oil and gas, primary metals, and transportation.
Certifications: ANSI Z87.1-2003. CSA Z94.3-2007.