Tryptic Soy Agar (TSA), USP, Hardy Diagnostics
Supplier: Hardy Diagnostics
U473 U60
89408-014PK 370.03 USD
89408-014 76299-204
Tryptic Soy Agar (TSA), USP, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For use as a general growth medium for the isolation and cultivation of microorganisms.
For use as a general growth medium for the isolation and cultivation of microorganisms. This medium is also recommended for use in the cultivation, storage, maintenance, and transportation of pure cultures of microorganisms.
Ordering information: Minimum order of 10 packs.