Denka-Seiken Salmonella Group O Antigens antisera, Hardy Diagnostics
294463 294449 294432 294425
10052-562EA 144.01 USD
10052-562 10052-588 10052-576 10052-586
Denka-Seiken Salmonella Group O Antigens antisera, Hardy Diagnostics
Determination of serotypes of Salmonella by slide agglutination and test tube agglutination.
These are liquid products, containing specific agglutinins for each O-grouping sera. H-sera and Vi serum, which are used for the serological study of Salmonella and for the determination of sero types of Salmonella by slide agglutination and test tube agglutination.