Microgen® Bioproducts Rotascreen® Latex Test, Hardy Diagnostics
Supplier: Hardy Diagnostics
M80
10062-750EA 246.29 USD
10062-750
Microgen® Bioproducts Rotascreen® Latex Test, Hardy Diagnostics
A rapid and simple latex agglutination test for the detection of Rotavirus antigen in fecal speciments.
A rapid and simple latex agglutination test for the detection of Rotavirus antigen in fecal speciments. Suitable for human or animal use. Specific and sensitive with a result in five minutes. No special equipment required.