Mycoplasma Broth, Hardy Diagnostics
Supplier: Hardy Diagnostics
U187
10053-810EA 367.28 USD
10053-810
Mycoplasma Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the detection of Mycoplasma spp
For the detection of Mycoplasma spp. associated with bovine mastitis.
associated with bovine mastitis.