Microgen® Bioproducts Microgen® Staph ID Panel, Hardy Diagnostics
Supplier: Hardy Diagnostics
MID69
10062-778EA 385.59 USD
10062-778
Microgen® Bioproducts Microgen® Staph ID Panel, Hardy Diagnostics
Microgen® Staph ID Panel, for the identification of Staphylococcus species.
  • Microwell format identification system
  • Minimizes handling and manipulation
  • Identify medically important members of the genus Staphylococcus
A microwell format identification system for Staphylococci of clinical, animal health and environmental importance
The test design minimizes handling and manipulation. Substrates selected specifically for Staphylococcus and related species. Simple and easy-to-use. Results in 24 hours.
The Microgen Staph-ID system employs 12 standardized biochemical substrates in microwells to identify medically important members of the genus Staphylococcus. It is capable of identifying 26 species of Staphylococcus, Kocuria, Kytococcus, and Micrococcus spp.
The Microgen Staph-ID system comprises a single microwell strip containing 12 standardized biochemical substrates which have been selected on the basis of extensive computer analysis of published databases for the identification of the genus Staphylococcus. The dehydrated substrates in each well are reconstituted with a suspension of the organism to be identified. The inoculum is prepared in the suspending medium provided. If the individual substrates are metabolized by the organism, a color change occurs during incubation or after addition of specific reagents.
Reagents required but not included in kit