Microgen® Bioproducts Microgen® ID Software on CD, Hardy Diagnostics
MID60
10032-746EA 28.16 USD
10032-746
Microgen® Bioproducts Microgen® ID Software on CD, Hardy Diagnostics
Software program providing users of Microgen® Identification Systems with an easy-to-use results analysis.
Software program providing users of Microgen® Identification Systems with an easy-to-use results analysis
After reading out the color reactions on the ID strip, an eight digit code is determined. This Octal Code is entered into the Microgen Identification System Software (MID60), which generates a report of the five most likely organisms in the selected database.
The software provides an identification based on probability, percent probability, and likelihood with an analysis of the quality of separation. Full definitions of these terms and an explanation of their usefulness and interpretation are provided with the software Help manual.
Provides probability and likelihood data.