Would you like to log in again?
Denka-Seiken Listeria Antisera , Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Antisera for the typing of Listeria Group O and H antigens.
Listeria Antisera consists of 8 O antisera: I/II, I, IV, V/VI, VI, VII, VIII, and IX; 2ml each vial and 4 H antisera: H-A, H-AB, H-C, H-D; 5ml each vial, by Denka-Seiken
Antisera for the typing of Listeria Group O and H antigens. Aids public health and food testing laboratories in identifying sources of infection/contamination to help prevent further cases of the disease. All items are also available separately in monovalent vial form.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
We are currently unable to calculate your contract price for this item. List price is being displayed temporarily. When your order is processed, you will be invoiced at your contract price even though list price is displayed now
Description Supplier No. VWR Catalog Number Unit Price Quantity
Listeria Antisera 294616 10052-570 Each (2ml) Retrieving
Product available on GSA Advantage®
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty