C. diff PRO, Hardy Diagnostics
Supplier: Hardy Diagnostics
Z120 Z120
10053-794EA 165.2 USD
CA10053-794 10053-794
C. diff PRO, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
Screen suspected Clostridium difficile isolates for L-proline aminopeptidase activity.
C. diff PRO™ Test Kit is recommended for use in screening suspected Clostridium difficile isolates for L-proline aminopeptidase activity and a lack of indole activity.
The test performs a proline separately from indole for a clear test interpretation. Rapid results in 5 minutes!