Rapid PNGase F Antibody Standard
Supplier: New England Biolabs
P6043S P6043S
103218-944EA 361.82 USD
CA103218-944 103218-944
Rapid PNGase F Antibody Standard
Antibodies
Rapid PNGase F Antibody Standard is a murine anti-MBP monoclonal antibody, isotype IgG2a. This antibody standard can be used as a positive control for Rapid PNGase F.
  • Contains typical IgG glycosylation in a conserved Asn (#295) of the Fc region
  • Optimal stability for up to 24 months