    Sort by:
  • Search Within Results
5-bromo-1-3-difluoro-2-methylbenzene
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Catalog Number: (103062-826)
Supplier:  Ark Pharm
Description:   5-Bromo-1,3-Difluoro-2-Methylbenzene, Purity: 95+%, CAS Number: 179617-08-4, Molecular Formula: C7H5BrF2, Molecular weight: 207.02, Appearance: Solid, Boiling Point: 176 deg C at 760 mmHg, Storage: 2-8 deg C, Size: 1g