    Sort by:
  • Search Within Results
methyl-6-chloro-5-methoxynicotinate
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Catalog Number: (103062-370)
Supplier:  Ark Pharm
Description:   METHYL 6-CHLORO-5-METHOXYNICOTINATE, Purity: 98%, CAS Number: 915107-31-2, Molecular Formula: C8H8ClNO3, Molecular Weight: 201.61, Appearance: Solid, Storage: 2-4 deg C, size: 250mg