    Sort by:
  • Search Within Results
You Searched for: "4-phenyl-1h-pyrazole"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Catalog Number: (103062-792)
Supplier:  Ark Pharm
Description:   4-Phenyl-1H-Pyrazole, Purity: 95+%, CAS Number: 10199-68-5, Molecular Formula: C9H8N2, Molecular weight: 144.17, Appearance: Solid, Boiling Point: 340.9 deg C at 760 mmHg, Storage: 2-8 deg C, Size: 1g