Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:
  • Search Within Results
You Searched for: "methyl-4-fluoro-1h-indole-5-carboxylate"
SearchPresentationType-VERTICAL
SearchResultCount:"1"
Sort by:
Supplier:  Ark Pharm
Description:   METHYL 4-FLUORO-1H-INDOLE-5-CARBOXYLATE, Purity: 98%, CAS Number: 1252782-43-6, Molecular Formula: C10H8FNO2, Molecular Weight: 193.17, Appearance: Solid, Storage: 2-8 deg C, size: 1g