    Sort by:
  • Search Within Results
You Searched for: "7-methoxy-3-4-dihydro-2h-benzo-b-1-4-oxazine"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Ark Pharm
Description:   7-Methoxy-3,4-Dihydro-2H-Benzo[B][1,4]Oxazine, Purity: 97%, CAS Number: 93735-22-9, Molecular Formula: C9H11NO2, Molecular weight: 165.19, Appearance: Solid, Boiling Point: 295.512 deg C at 760 mmHg, Storage: 2-8 deg C, Size: 250mg