    Sort by:
  • Search Within Results
You Searched for: "iodomethyl-cyclohexane"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Catalog Number: (103062-390)
Supplier:  Ark Pharm
Description:   (IODOMETHYL)CYCLOHEXANE, Purity: 95+%, CAS Number: 5469-33-0, Molecular Formula: C7H13I, Molecular Weight: 224.08, Appearance: Liquid, Boiling Point: 213.3 deg C at 760 mmHg, Storage: 2-4 deg C, size: 1g