You Searched For: 7-Bromo-2-methyl-1,2,3,4-tetrahydroisoquinoline
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Ambeed
Description: 7-Bromo-2-methyl-1,2,3,4-tetrahydroisoquinoline, Purity: 95%, CAS Number: 258515-54-7, Appearance: Liquid, Storage: Keep in dark place, Inert atmosphere, 2-8C, Size: 250MG