Proheat® Heat Guns, Master Appliance
Supplier: Master Appliance
Proheat®
PH-1400K PH-1200 PH-1100 PH-1600 PH-1400
94025-812EA 403.61 USD
94025-812 75845-496 94025-810 75845-494 75845-502
Proheat® Heat Guns, Master Appliance
Heaters Hot Air Dryers
Use these Proheat guns for laboratory work, activate adhesives, bend plastics, dry filler material, shrink tubing and packaging, stretch vinyls, strip paint and more.
  • Compact, lightweight, easy to use
  • Dual temperature settings
  • Balanced ergonomic design prevents operator fatigue
  • Protective hand/switch guard and hanging loop
Electronic closed loop temperature control system maintains constant temperature and protects unit from overheating. Additional features include dial-In, LCD, and surface temperature control.
Certifications: UL and CUL listed or CSA certified, made in USA
Accessories information: Accessories available separately.