Difco™ Dextrose Broth, BD
263100 263100
90003-732EA 159.17 USD
90003-732 CA90003-732
Difco™ Dextrose Broth, BD
Dextrose Broth is used for cultivating fastidious microorganisms and for detecting gas from enteric bacilli.
  • Low agar concentration
  • Beef extract and peptones provide essential nutrients
  • Dextrose is a carbon source
  • Increased concentration of dextrose
Supplementation with 5% blood provides additional growth factors for fastidious microorganisms.