Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Density Meters, DMA Generation M, Anton Paar

Supplier: Anton Paar
153058 153059 153060
The DMA 5000 M is the most accurate density meter on the market and is ideal for numerous fields of application and successful continuous operation in the laboratory.

  • 10.4" touchscreen
  • Applies state-of-the-art PCAP technology
  • Provides a premium user experience
  • FillingCheck™
  • Detects filling errors and gas bubbles in the sample
  • Generates and records filling warnings
  • U-View™
  • Displays live images of the oscillating U-tube sensor and the entire filled-in sample
  • Relevant images with the sample fully and clearly visible are stored in the data memory
  • Later review of these images guarantees completely transparent measurement processes
  • Particularly when using automatic sampling systems, U-View™ and FillingCheck™ supervise the entire measurement sequence and allow later verification of the results
  • ThermoBalance™
  • Eliminates all measurement drifts caused by temperature stress due to temperature changes or sample temperatures greatly different from the measuring temperature
  • Allows accurate and convenient density measurements in a wide temperature range
  • Only one adjustment at one temperature required
  • Reference oscillator

First-class digital density measurement provided by the robust DMA Generation M density meters is applied in industrial quality control, in research, at authorities and standards organizations

This series of density meters - completed by DMA 4100 M and DMA 4500 M - provides unparalleled ease-of-use at an attractive price and ensures complete transparency and traceability of the sample filling and measurement process. Automatic viscosity correction. Across the samples' entire viscosity range. Eliminates viscosity-related errors. No viscosity standards required.

Be sure of your results. Fast, reliable measurement results at the highest accuracy (up to 0.000005 g/cm³). Only 1 mL of sample required. Preset measuring methods for different applications. Integrated conversion tables for simple density and concentration measurement. Different user functions applicable: tables, polynomials, formulas, linear functions and constants.

Full QM compliance. GMP/GLP and 21 CFR Part 11 compatibility. Audit Trail. Password protection with three user levels. Forgery-proof data export. Different security levels and increased security settings. Electronic signature.

Integrated tables and Functions: Alcohol tables, Extract/sugar tables, API functions, Acid/base tables, 20 freely programmable tables/user functions (tables, polynomials, formulas, linear functions and constants).
Wetted parts include PTFE, borosilicate glass.

Certifications: UL and CE Certified.

Ordering information: Optional Accessories include Printers, Density Standards and Pharmaceutical Qualifications.




 

ORDER

Expand All / Collapse All
Description Accuracy Measurement Range Repeatability (Std. Dev.) Supplier No. VWR Catalog Number Unit Price Quantity
Density Meter DMA 5000M Density 0.000005 g/cm³;
Temperature 0.01 °C/0.02 °F
0 to 3 g/cm3 Density 0.000001 g/cm³;
Temperature 0.001°C/0.002°F
183345 75801-606 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Product available on GSA Advantage®
Density Meter DMA 4500M Density 0.00005 g/cm³ ;
Temperature 0.03 °C/0.05 °F
0 to 3 g/cm3 Density 0.00001 g/cm³;
Temperature 0.01 °C/0.02 °F
176803 75801-604 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Product available on GSA Advantage®
Density Meter DMA 4100M Density 0.0001 g/cm³;
Temperature 0.05 °C/0.09 °F
0 to 3 g/cm3 Density 0.00005 g/cm³;
Temperature 0.02 °C/0.04 °F
163378 75801-602 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Product available on GSA Advantage®
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Temperature Range (°C) 0°C to 100°C (32°F to 212°F)
Sample Size approx. 1 mL (Minimum)
Interfaces 4 x USB, 2 x S-BUS, RS-232, CAN, VGA, Ethernet
Memory 1000 measurement results (optional ring memory)
Electrical AC 100 to 240V; 50 to 60Hz; 190VA
Weight 22.5 kg (49.6 lbs)
Dimensions 495 x 330 x 230 mm (19.5 x 13 x 9.1")

ACCESSORIES

Expand All / Collapse All

Accessories
Description Details Supplier No. VWR Catalog Number Unit Price Quantity
Density Standard, Dodecane, 10 mL 302956 75801-612 Each (10ml) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Density Standard, Water, 5 x 10 mL 96044 75801-610 Each(10ml) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Pharma Qualification Documentation Price for paperwork only; Labor and travel charges will apply and are only quoted through Anton Paar 156610 75801-614 Each Retrieving
Pharma Qualification Smart Documentation Price for paperwork only; Labor and travel charges will apply and are only quoted through Anton Paar 102160 75801-616 Each Retrieving
Printer CBM-910II with Cable 64434 75801-608 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty