Formaldehyde 37% (w/w), Surgipath® Histological Fixative

Supplier: Leica Biosystems
Danger

Synonyms: MethylaldehydeFormolFormalinParaformaldehyde aqueous solution

Surgipath®
3800750
75810-340CS 149.68 USD
75810-340
Formaldehyde 37% (w/w), Surgipath® Histological Fixative
Formaldehyde

This product is made available for those who desire to create their own buffered formulas or where higher than 10% product ratio to water is required.


  • Used to create buffered formulas
  • For use with 10% product to water ratio


40% by volume, 37% by weight, stabilized with methanol to inhibit polymerzation.

Formula: CH₂O
MW: 30.03 g/mol
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 2209
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR