Formalin concentrate 10%, Surgipath® fixative reagent, buffered
Supplier: Leica Biosystems
|
Danger
|
Synonyms:
Methylaldehyde
, Formol
, Paraformaldehyde aqueous solution
, Formaldehyde
3800540
75810-336EA
54.54
USD
75810-336
Formalin concentrate 10%, Surgipath® fixative reagent, buffered
Formalin
Buffered Formalin Concentrate 5 mixes one part (one gallon) to four parts (four gallons) water to make a five gallon batch of 10% Neutral Buffered Formalin.
- 1:4 to make 5 gallons
- 10% neutral buffered formalin
Formula:
CH₂O MW: 30.03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003274 CAS Number: 50-00-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...